Assay Details
Gene Symbol: Pacsin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289677.1 | NP_001276606.1 | 6 - 7 | 994 | 68 | |
NM_001289678.1 | NP_001276607.1 | 7 - 8 | 993 | 68 | ||
NM_028733.4 | NP_083009.1 | 7 - 8 | 1123 | 68 | ||
NM_030880.3 | NP_112019.2 | 7 - 8 | 989 | 68 | ||
GenBank mRNA | AF149824.1 | - | 7 - 8 | 907 | 68 | |
AF242531.1 | - | 6 - 7 | 892 | 68 | ||
AK049166.1 | - | 7 - 8 | 1124 | 68 | ||
AK078060.1 | - | 6 - 7 | 1000 | 68 | ||
AK141063.1 | - | 7 - 8 | 966 | 68 | ||
BC003884.1 | - | 7 - 8 | 930 | 3497435 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
