Assay Details
Gene Symbol: Cnpy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF361644.1 | - | 5 - 6 | 704 | 104 | |
AK005532.1 | - | 5 - 6 | 687 | 104 | ||
AK031742.1 | - | 5 - 6 | 542 | 104 | ||
AK049358.1 | - | 5 - 6 | 690 | 104 | ||
AK082340.1 | - | 3 - 4 | 470 | 104 | ||
AK082749.1 | - | 3 - 4 | 306 | 104 | ||
AK085617.1 | - | 5 - 6 | 697 | 104 | ||
AK086940.1 | - | 5 - 6 | 637 | 104 | ||
AK143428.1 | - | 5 - 6 | 680 | 104 | ||
AK220209.1 | - | 5 - 6 | 514 | 104 | ||
BC013549.1 | - | 5 - 6 | 706 | 4222133 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31208746 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31208748 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31208747 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P498403 | AF361644.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P498403 | AF361644.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925577 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 126391 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 80976 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 626193 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508820 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 106131 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906352 | AK028062.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906352 | AK075613.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906352 | BC011343.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906352 | NM_144846.4 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 503228 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75048 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 67541 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 16514 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134107 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P498403 | AF361644.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P498403 | AF361644.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208731 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208735 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208741 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208744 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208748 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208758 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208766 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208756 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208752 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208759 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208737 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208747 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208745 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208750 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208732 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208746 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208740 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208753 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208751 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208749 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208764 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208761 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208754 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08790698 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08790704 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134107 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03325360 | - | - |
Whole Mouse Genome G4122A | A_51_P498403 | AF361644.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P498403 | AF361644.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105164 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03325360 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08790698 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134107 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13294920 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13398221 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13398231 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31208748 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31208747 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31208746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208637 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208788 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208656 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208678 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208615 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208620 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208612 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208686 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208679 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208713 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208706 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13398218 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208732 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208751 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208707 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208744 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208730 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208762 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208672 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208725 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13558935 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208705 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208709 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208721 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208735 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208708 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208745 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208766 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208676 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13182134 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208764 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13404963 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208737 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208717 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13372180 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208758 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208724 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208756 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208711 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208748 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208694 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208704 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208761 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208731 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208749 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13447163 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208754 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208670 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208722 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208720 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208726 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208675 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208698 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208752 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208728 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208714 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208712 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208673 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208701 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208759 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208741 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31208696 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10451464 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10451459 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10451463 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10451461 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10451462 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10451460 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1426479_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10451458 | - | - |
Mouse Genome 430 2.0 Array | 1426479_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1426479_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72029_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72029_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1700020661.mm.1 | TC1700002189.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700020661.mm.1 | NM_028065.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700020662.mm.1 | TC1700002189.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700020662.mm.1 | NM_028065.4 | Refseq |
Mouse Expression Set 430 | 1426479_PM_a_at | - | - |
Mouse Expression Set 430 | 1426479_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1426479_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1426479_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426479_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426479_PM_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2100121 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2100121 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3840519 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003840519 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002100121 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244634 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223615 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1223615 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1244634 | - | - |
mouse-6 v1.1 (Illumina) | scl49852.7_79-S | - | - |
mouse-6 v1.1 (Illumina) | scl0001755.1_21-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2100121 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3840519 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001755.1_21-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl49852.7_79-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3840519 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2100121 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1244634 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244634 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl49852.7_79-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001755.1_21-S | - | - |
