Assay Details
Gene Symbol: P2ry2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK155521.1 | - | 1 - 2 | 326 | 65 | |
AK155562.1 | - | 1 - 2 | 326 | 65 | ||
AK156185.1 | - | 2 - 3 | 278 | 65 | ||
BC006613.1 | - | 2 - 3 | 468 | 3967332 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK155521.1, AK155562.1, AK156185.1, BC006613.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
