Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700012496.mm.1 |
TC0700001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012496.mm.1 |
NM_001286689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012496.mm.1 |
NM_001286690.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012496.mm.1 |
NM_011644.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012496.mm.1 |
NR_104582.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012499.mm.1 |
TC0700001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012499.mm.1 |
NM_001286689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012499.mm.1 |
NM_001286690.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012499.mm.1 |
NM_011644.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012499.mm.1 |
NR_104582.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012493.mm.1 |
TC0700001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012493.mm.1 |
NM_001286689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012493.mm.1 |
NM_001286690.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012493.mm.1 |
NM_011644.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012493.mm.1 |
NR_104582.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012495.mm.1 |
TC0700001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012495.mm.1 |
NM_001286689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012495.mm.1 |
NM_001286690.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012495.mm.1 |
NM_011644.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012495.mm.1 |
NR_104582.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012492.mm.1 |
TC0700001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012492.mm.1 |
NM_001286689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012492.mm.1 |
NM_001286690.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012492.mm.1 |
NM_011644.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012492.mm.1 |
NR_104582.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012506.mm.1 |
TC0700001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012506.mm.1 |
NM_001286689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012506.mm.1 |
NM_001286690.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012506.mm.1 |
NM_011644.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012506.mm.1 |
NR_104582.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012514.mm.1 |
TC0700001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012514.mm.1 |
NM_001286689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012514.mm.1 |
NM_001286690.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012517.mm.1 |
TC0700001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012517.mm.1 |
NM_001286689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012517.mm.1 |
NM_001286690.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012520.mm.1 |
TC0700001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012520.mm.1 |
NM_001286689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012520.mm.1 |
NM_001286690.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012503.mm.1 |
TC0700001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012503.mm.1 |
NM_001286689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012503.mm.1 |
NM_001286690.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012503.mm.1 |
NM_011644.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012503.mm.1 |
NR_104582.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012513.mm.1 |
TC0700001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012513.mm.1 |
NM_001286689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012513.mm.1 |
NM_001286690.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012515.mm.1 |
TC0700001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012515.mm.1 |
NM_001286689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012515.mm.1 |
NM_001286690.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012518.mm.1 |
TC0700001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012518.mm.1 |
NM_001286689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012518.mm.1 |
NM_001286690.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012500.mm.1 |
TC0700001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012500.mm.1 |
NM_001286689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012500.mm.1 |
NM_001286690.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012500.mm.1 |
NM_011644.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012500.mm.1 |
NR_104582.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012497.mm.1 |
TC0700001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012497.mm.1 |
NM_001286689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012497.mm.1 |
NM_001286690.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012497.mm.1 |
NM_011644.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012497.mm.1 |
NR_104582.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012498.mm.1 |
TC0700001446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700012498.mm.1 |
NM_001286689.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012498.mm.1 |
NM_001286690.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012498.mm.1 |
NM_011644.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700012498.mm.1 |
NR_104582.1 |
Refseq |