Assay Details
Gene Symbol: Dennd5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021494.1 | NP_067469.1 | 11 - 12 | 2518 | 69 | |
GenBank mRNA | AJ245569.1 | - | 10 - 11 | 2424 | 69 | |
AK084641.1 | - | 5 - 6 | 1997 | 69 | ||
BC022119.1 | - | 7 - 8 | 1171 | 5067385 | 69 | |
BC060230.1 | - | 11 - 12 | 2518 | 6840562 | 69 | |
L40894.1 | - | 2 - 3 | 166 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
