Assay Details
Gene Symbol: Arhgap17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122641.1 | NP_001116113.1 | 16 - 17 | 1597 | 84 | |
NM_001122642.1 | NP_001116114.1 | 16 - 17 | 1597 | 84 | ||
NM_001122643.1 | NP_001116115.1 | 16 - 17 | 1597 | 84 | ||
GenBank mRNA | AK036468.1 | - | 16 - 17 | 1534 | 84 | |
AK036617.1 | - | 16 - 17 | 1534 | 84 | ||
AK149986.1 | - | 16 - 17 | 1569 | 84 | ||
AK152266.1 | - | 16 - 17 | 1534 | 84 | ||
BC003259.1 | - | 16 - 17 | 1545 | 3500822 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
