Assay Details
Gene Symbol: Arhgap17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122643.1 | NP_001116115.1 | 19 - 20 | 2334 | 101 | |
GenBank mRNA | AK046411.1 | - | 19 - 20 | 2476 | 101 | |
AK149986.1 | - | 19 - 20 | 2306 | 101 | ||
BC003259.1 | - | 19 - 20 | 2273 | 3500822 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
