Assay Details
Gene Symbol: Cdhr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114322.1 | NP_001107794.1 | 5 - 6 | 581 | 90 | |
NM_028069.3 | NP_082345.1 | 5 - 6 | 581 | 90 | ||
GenBank mRNA | AF462391.1 | - | 5 - 6 | 570 | 90 | |
AK007988.1 | - | 5 - 6 | 570 | 90 | ||
AK027913.1 | - | 5 - 6 | 582 | 90 | ||
BB609487.1 | - | 5 - 6 | 582 | 90 | ||
BC043336.1 | - | 5 - 6 | 546 | 5042289 | 90 | |
BC054469.1 | - | 5 - 6 | 566 | 4225164 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
