Assay Details
Gene Symbol: Gpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145820.1 | NP_001139292.1 | 13 - 14 | 2080 | 92 | |
NM_010274.3 | NP_034404.3 | 13 - 14 | 1955 | 92 | ||
GenBank mRNA | AK034353.1 | - | 13 - 14 | 1864 | 92 | |
AK144716.1 | - | 13 - 14 | 2081 | 92 | ||
AK151189.1 | - | 13 - 14 | 1868 | 92 | ||
AK167152.1 | - | 13 - 14 | 1893 | 92 | ||
BC021359.1 | - | 13 - 14 | 2049 | 5012142 | 92 | |
D50430.1 | - | 13 - 14 | 1857 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
