Assay Details
Gene Symbol: Gpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145820.1 | NP_001139292.1 | 7 - 8 | 1141 | 55 | |
NM_010274.3 | NP_034404.3 | 7 - 8 | 1016 | 55 | ||
GenBank mRNA | AK034353.1 | - | 7 - 8 | 925 | 55 | |
AK144716.1 | - | 7 - 8 | 1142 | 55 | ||
AK151189.1 | - | 7 - 8 | 928 | 55 | ||
AK167152.1 | - | 7 - 8 | 954 | 55 | ||
BC021359.1 | - | 7 - 8 | 1110 | 5012142 | 55 | |
D50430.1 | - | 7 - 8 | 918 | 55 | ||
U60987.1 | - | 7 - 8 | 952 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
