Assay Details
Gene Symbol: Sec23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281816.1 | NP_001268745.1 | 1 - 2 | 107 | 80 | |
GenBank mRNA | AK029661.1 | - | 1 - 2 | 108 | 80 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK029661.1, NM_001281816.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
