Assay Details
Gene Symbol: Xrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011917.2 | NP_036047.2 | 5 - 6 | 549 | 121 | |
GenBank mRNA | AK004800.1 | - | 5 - 6 | 524 | 121 | |
AK011711.1 | - | 5 - 6 | 547 | 121 | ||
AK031247.1 | - | 5 - 6 | 592 | 121 | ||
AK053643.1 | - | 5 - 6 | 542 | 121 | ||
AK133654.1 | - | 5 - 6 | 531 | 121 | ||
AK167019.1 | - | 5 - 6 | 569 | 121 | ||
AK168037.1 | - | 5 - 6 | 595 | 121 | ||
BC052823.1 | - | 5 - 6 | 514 | 121 | ||
BC054743.1 | - | 5 - 6 | 549 | 6400974 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
