Assay Details
Gene Symbol: Polr3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164096.1 | NP_001157568.1 | 7 - 8 | 643 | 124 | |
NM_025298.3 | NP_079574.2 | 8 - 9 | 721 | 124 | ||
GenBank mRNA | AK005034.1 | - | 7 - 8 | 644 | 124 | |
AK012184.1 | - | 8 - 9 | 681 | 124 | ||
AK144721.1 | - | 8 - 9 | 684 | 125 | ||
AK146279.1 | - | 8 - 9 | 678 | 124 | ||
AK159723.1 | - | 8 - 9 | 678 | 124 | ||
BC028924.1 | - | 8 - 9 | 642 | 124 | ||
BC037637.1 | - | 8 - 9 | 671 | 5010681 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
