Assay Details
Gene Symbol: Fuom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286217.1 | NP_001273146.1 | 4 - 5 | 375 | 117 | |
NM_001286218.1 | NP_001273147.1 | 3 - 4 | 304 | 117 | ||
NM_026928.3 | NP_081204.2 | 4 - 5 | 375 | 117 | ||
NR_104415.1 | 4 - 5 | 375 | 117 | |||
GenBank mRNA | AA403787.1 | - | 3 - 4 | 235 | 117 | |
AK042452.1 | - | 338 | 117 | |||
AK044731.1 | - | 4 - 5 | 340 | 117 | ||
BC028662.1 | - | 4 - 5 | 336 | 1332340 | 117 | |
CK618091.1 | - | 4 - 5 | 310 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987231 | AK044731.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987231 | AK079410.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987231 | AK163708.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987231 | BC028662.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987231 | NM_026928.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987226 | AK079410.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P466701 | AK044731.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P466701 | BC028662.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P466701 | NM_026928.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P466701 | AK007494.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P466701 | AK044731.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P466701 | BC028662.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P466701 | NM_026928.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P466701 | AK007494.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987222 | AK042452.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 547681 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 587791 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 300643 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 356212 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116541 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987231 | AK044731.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987231 | AK079410.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987231 | AK163708.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987231 | BC028662.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987231 | NM_026928.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P466701 | AK044731.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P466701 | BC028662.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P466701 | NM_026928.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P466701 | AK007494.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P466701 | AK044731.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P466701 | BC028662.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P466701 | NM_026928.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P466701 | AK007494.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582255 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582247 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582238 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582270 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582274 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582271 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582278 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582254 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582261 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582259 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582272 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582249 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582266 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582257 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582240 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582275 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582245 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582251 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582276 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582264 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582256 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582263 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582237 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582242 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582243 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582258 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582248 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25582268 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116541 | - | - |
Whole Mouse Genome G4122A | A_52_P466701 | AK044731.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P466701 | BC028662.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P466701 | NM_026928.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P466701 | AK007494.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P466701 | AK044731.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P466701 | BC028662.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P466701 | NM_026928.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P466701 | AK007494.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115271 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116541 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06089532 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06089525 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06658704 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06089529 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06089531 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06089527 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06658702 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06089530 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06089524 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582230 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582212 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582204 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582201 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582317 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582202 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582209 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582231 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25582221 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06658704 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06197427 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582248 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582245 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582272 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06089527 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582243 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06089532 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582238 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582269 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582247 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582240 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582259 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582268 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06089529 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582235 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582270 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05966099 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582278 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582274 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06552352 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582251 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582271 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582252 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582234 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06089524 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582275 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582263 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582257 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582249 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25582255 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10568932 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10568934 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10568936 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10568938 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10568935 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10568937 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1452626_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10568930 | - | - |
Mouse Genome 430 2.0 Array | 1452626_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1452626_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69064_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69064_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700037740.mm.1 | TC0700004493.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700037740.mm.1 | NR_104415.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700037742.mm.1 | TC0700004493.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700037742.mm.1 | NM_001286217.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700037742.mm.1 | NM_001286218.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700037742.mm.1 | NM_026928.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700037742.mm.1 | NR_104415.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700037733.mm.1 | TC0700004493.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700037733.mm.1 | NM_001286217.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700037733.mm.1 | NM_001286218.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700037733.mm.1 | NM_026928.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700037733.mm.1 | NR_104415.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700037732.mm.1 | TC0700004493.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700037732.mm.1 | NM_001286217.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700037732.mm.1 | NM_001286218.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700037732.mm.1 | NM_026928.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700037732.mm.1 | NR_104415.1 | Refseq |
Mouse Expression Set 430 | 1452626_a_at | - | - |
Mouse Expression Set 430 | 1452626_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1452626_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1452626_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452626_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452626_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4010520 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4010520 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 104810411 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6900041 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004010520 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006900041 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1251065 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1258535 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1251065 | - | - |
mouse-6 v1.1 (Illumina) | scl23951.18_468-S | - | - |
mouse-6 v1.1 (Illumina) | scl069064.1_23-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 104810411 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6900041 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4010520 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl23951.18_468-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl069064.1_23-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4010520 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6900041 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104810411 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1251065 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1251930 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1258535 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1251065 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1251930 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl069064.1_23-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl23951.18_468-S | - | - |
