Assay Details
Gene Symbol: Fuom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104415.1 | 5 - 6 | 527 | 67 | ||
GenBank mRNA | AK044731.1 | - | 5 - 6 | 492 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
