Assay Details
Gene Symbol: Slc5a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177624.1 | NP_001171095.1 | 10 - 11 | 1528 | 67 | |
GenBank mRNA | AK128951.1 | - | 10 - 11 | 1520 | 67 | |
AK144004.1 | - | 10 - 11 | 1461 | 67 | ||
AK165419.1 | - | 10 - 11 | 1521 | 67 | ||
AY964639.1 | - | 10 - 11 | 1231 | 67 | ||
BC121791.1 | - | 10 - 11 | 1224 | 67 | ||
BC141255.1 | - | 10 - 11 | 1456 | 9055867 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
