Assay Details
Gene Symbol: Chrna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007389.5 | NP_031415.2 | 2 - 3 | 243 | 75 | |
GenBank mRNA | AK029177.1 | - | 2 - 3 | 212 | 75 | |
AK132589.1 | - | 2 - 3 | 183 | 75 | ||
BC125439.1 | - | 2 - 3 | 243 | 40129954 | 75 | |
BC137678.1 | - | 2 - 3 | 243 | 8860697 | 75 | |
EF520684.1 | - | 1 - 2 | 118 | 75 | ||
X03986.1 | - | 2 - 3 | 200 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
