Assay Details
Gene Symbol: Mki67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081117.2 | NP_001074586.2 | 9 - 10 | 2145 | 89 | |
GenBank mRNA | AK052772.1 | - | 9 - 10 | 2065 | 89 | |
AK082099.1 | - | 9 - 10 | 2064 | 89 | ||
AK143677.1 | - | 9 - 10 | 2184 | 89 | ||
X82786.1 | - | 9 - 10 | 2253 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
