Assay Details
Gene Symbol: Rgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009059.2 | NP_033085.2 | 12 - 13 | 1573 | 68 | |
GenBank mRNA | AK144440.1 | - | 12 - 13 | 1575 | 68 | |
BC068121.1 | - | 12 - 13 | 1573 | 6415928 | 68 | |
KJ650210.1 | - | 11 - 12 | 1381 | 68 | ||
U54639.1 | - | 12 - 13 | 1502 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
