Assay Details
Gene Symbol: Rgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009059.2 | NP_033085.2 | 14 - 15 | 1792 | 82 | |
GenBank mRNA | BC068121.1 | - | 14 - 15 | 1792 | 6415928 | 82 |
KJ650210.1 | - | 13 - 14 | 1600 | 82 | ||
U54639.1 | - | 14 - 15 | 1721 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
