Assay Details
Gene Symbol: Rgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009059.2 | NP_033085.2 | 2 - 3 | 354 | 78 | |
GenBank mRNA | AK144440.1 | - | 2 - 3 | 353 | 78 | |
BC068121.1 | - | 2 - 3 | 354 | 6415928 | 78 | |
KJ650210.1 | - | 1 - 2 | 162 | 78 | ||
U54639.1 | - | 2 - 3 | 283 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
