Assay Details
Gene Symbol: Myoz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021508.3 | NP_067483.1 | 5 - 6 | 787 | 80 | |
GenBank mRNA | AJ005620.1 | - | 5 - 6 | 727 | 80 | |
AK009077.1 | - | 5 - 6 | 751 | 80 | ||
AK041896.1 | - | 5 - 6 | 743 | 80 | ||
AY013298.1 | - | 5 - 6 | 705 | 80 | ||
BC024660.1 | - | 5 - 6 | 843 | 3967841 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29448045 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29448047 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29448046 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29448043 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29448048 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29448044 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29448049 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P379750 | AY013298.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P379750 | BC024660.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P379750 | AJ005620.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P379750 | AK009077.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P379750 | AK041896.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P379750 | AY013298.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P379750 | BC024660.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P379750 | AJ005620.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P379750 | AK041896.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P379750 | AK009077.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 319686 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392615 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 957580 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 382990 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 78922 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164052 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P379750 | AY013298.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P379750 | BC024660.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P379750 | AJ005620.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P379750 | AK009077.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P379750 | AK041896.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P379750 | AY013298.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P379750 | BC024660.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P379750 | AJ005620.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P379750 | AK009077.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P379750 | AK041896.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29448151 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29448146 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29448124 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29448120 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29448138 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29448126 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29448142 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29448130 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29448137 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29448118 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29448127 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29448131 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29448139 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29448149 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29448117 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29448150 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29448136 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29448129 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29448133 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29448122 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164052 | - | - |
Whole Mouse Genome G4122A | A_52_P305767 | AY013298.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P305767 | BC024660.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P305767 | AJ005620.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P305767 | AK009077.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P305767 | AK041896.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P379750 | AY013298.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P379750 | BC024660.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P379750 | AJ005620.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P379750 | AK009077.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P379750 | AK041896.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P379750 | AY013298.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P379750 | BC024660.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P379750 | AJ005620.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P379750 | AK009077.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P379750 | AK041896.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114388 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164052 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11463106 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11463103 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11463110 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11463117 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11463120 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29448049 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29448048 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29448045 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29448043 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29448047 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29448044 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29448046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448067 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448212 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448208 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448209 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448195 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448136 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448198 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448201 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29448127 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448136 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11531506 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448131 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448110 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448052 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11647218 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11463109 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448102 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11124336 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11531503 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448048 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11818714 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11463112 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448098 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448150 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11531497 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448051 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448105 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448142 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11463115 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448042 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448124 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11172780 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448039 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448032 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448036 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448083 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11814712 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448151 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448107 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448111 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11581747 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448045 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448038 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448130 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448086 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448120 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448099 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448144 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448113 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448115 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11814717 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448118 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448037 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11531502 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448127 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448078 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11463110 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448096 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448093 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11785238 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448092 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448114 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448095 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11531515 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11814716 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11327478 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11531492 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448080 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29448034 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10417935 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10417937 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10417939 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10417938 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10417936 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10417934 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1448636_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1460202_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10417933 | - | - |
Mouse Genome 430 2.0 Array | 1460202_at | - | - |
Mouse Genome 430 2.0 Array | 1448636_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1460202_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1448636_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 59011_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 59011_PM_at | - | - |
Mouse Expression Set 430 | 1448636_at | - | - |
Mouse Expression Set 430 | 1460202_PM_at | - | - |
Mouse Expression Set 430 | 1460202_at | - | - |
Mouse Expression Set 430 | 1448636_PM_at | - | - |
Murine Genome U74v2 Set | 104555_PM_at | - | - |
Murine Genome U74v2 Set | 104555_at | - | - |
Mouse Genome 430A 2.0 Array | 1460202_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1448636_at | - | - |
Mouse Genome 430A 2.0 Array | 1448636_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1460202_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1460202_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448636_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1460202_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448636_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 923071 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2760670 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2760670 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002760670 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2615250 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2615250 | - | - |
mouse-6 v1.1 (Illumina) | scl45844.6.1_21-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2760670 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl45844.6.1_21-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2760670 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2615250 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2615250 | - | - |
