Assay Details
Gene Symbol: Scai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178778.3 | NP_848893.2 | 1 - 2 | 136 | 112 | |
GenBank mRNA | AK044772.1 | - | 1 - 2 | 132 | 112 | |
AK083280.1 | - | 1 - 2 | 138 | 112 | ||
AK163712.1 | - | 1 - 2 | 140 | 112 | ||
BC132485.1 | - | 1 - 2 | 111 | 40130762 | 112 | |
BY783760.1 | - | 1 - 2 | 140 | 112 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK044772.1, AK083280.1, AK163712.1, BC132485.1, NM_178778.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
