Assay Details
Gene Symbol: Mtmr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016985.2 | NP_058681.1 | 10 - 11 | 1409 | 144 | |
GenBank mRNA | AF073997.1 | - | 10 - 11 | 1382 | 144 | |
AK048714.1 | - | 2 - 3 | 662 | 144 | ||
AK051390.1 | - | 3 - 4 | 407 | 144 | ||
BC056376.1 | - | 10 - 11 | 1409 | 6849757 | 144 | |
BC057337.1 | - | 10 - 11 | 1403 | 6825916 | 144 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
