Assay Details
Gene Symbol: Mtmr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016985.2 | NP_058681.1 | 4 - 5 | 591 | 83 | |
GenBank mRNA | AF073997.1 | - | 4 - 5 | 564 | 83 | |
AY099894.1 | - | 4 - 5 | 289 | 83 | ||
AY099895.1 | - | 5 - 6 | 316 | 83 | ||
AY099896.1 | - | 6 - 7 | 367 | 83 | ||
AY099897.1 | - | 4 - 5 | 316 | 83 | ||
AY099898.1 | - | 5 - 6 | 340 | 83 | ||
BC056376.1 | - | 4 - 5 | 591 | 6849757 | 83 | |
BC057337.1 | - | 4 - 5 | 585 | 6825916 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
