Assay Details
Gene Symbol: Ap3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009680.3 | NP_033810.2 | 21 - 22 | 2646 | 77 | |
GenBank mRNA | AF103809.2 | - | 21 - 22 | 2583 | 77 | |
AK163996.1 | - | 6 - 7 | 704 | 77 | ||
AK203980.1 | - | 1 - 2 | 82 | 77 | ||
BC015068.1 | - | 21 - 22 | 2652 | 4018563 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
