Assay Details
Gene Symbol: Hmgcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008255.2 | NP_032281.2 | 15 - 16 | 2129 | 85 | |
GenBank mRNA | AK079302.1 | - | 3 - 4 | 328 | 85 | |
AK159899.1 | - | 14 - 15 | 2017 | 85 | ||
BC019782.1 | - | 14 - 15 | 2005 | 85 | ||
BC059873.1 | - | 6 - 7 | 810 | 85 | ||
BC085083.1 | - | 15 - 16 | 2041 | 5320486 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
