Assay Details
Gene Symbol: Fcho2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172591.3 | NP_766179.2 | 22 - 23 | 2324 | 98 | |
GenBank mRNA | AK031041.1 | - | 22 - 23 | 2327 | 98 | |
AK142282.1 | - | 22 - 23 | 2010 | 98 | ||
AK161093.1 | - | 3 - 4 | 1057 | 98 | ||
AK164135.1 | - | 23 - 24 | 2182 | 98 | ||
AK169439.1 | - | 6 - 7 | 1746 | 98 | ||
BC052456.1 | - | 22 - 23 | 2021 | 6830607 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
