Assay Details
Gene Symbol: Fcho2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172591.3 | NP_766179.2 | 5 - 6 | 837 | 136 | |
GenBank mRNA | AK031041.1 | - | 5 - 6 | 840 | 136 | |
AK142282.1 | - | 5 - 6 | 523 | 136 | ||
AK146566.1 | - | 5 - 6 | 571 | 136 | ||
AK164135.1 | - | 5 - 6 | 583 | 136 | ||
BC019933.1 | - | 6 - 7 | 573 | 136 | ||
BC047122.1 | - | 6 - 7 | 573 | 136 | ||
BC052456.1 | - | 5 - 6 | 534 | 6830607 | 136 | |
BC053718.1 | - | 5 - 6 | 553 | 136 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK164135.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
