Assay Details
Gene Symbol: Rad50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009012.2 | NP_033038.2 | 13 - 14 | 2471 | 122 | |
GenBank mRNA | AK037641.1 | - | 12 - 13 | 2314 | 122 | |
U66887.1 | - | 13 - 14 | 2449 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
