Assay Details
Gene Symbol: Rad50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009012.2 | NP_033038.2 | 22 - 23 | 3734 | 82 | |
GenBank mRNA | AK132949.1 | - | 2 - 3 | 574 | 82 | |
U66887.1 | - | 22 - 23 | 3712 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
