Assay Details
Gene Symbol: Hif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010431.2 | NP_034561.2 | 3 - 4 | 791 | 141 | |
GenBank mRNA | AF003695.1 | - | 3 - 4 | 625 | 141 | |
AK017853.1 | - | 3 - 4 | 652 | 141 | ||
AK034087.1 | - | 3 - 4 | 757 | 141 | ||
AK076395.1 | - | 1 - 2 | 79 | 141 | ||
AK138944.1 | - | 3 - 4 | 658 | 141 | ||
AK150367.1 | - | 3 - 4 | 658 | 141 | ||
BC026139.1 | - | 3 - 4 | 643 | 4019056 | 141 | |
X95580.1 | - | 3 - 4 | 457 | 141 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
