Assay Details
Gene Symbol: Prpf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AI790078.1 | - | 2 - 3 | 274 | 89 | |
AK154170.1 | - | 9 - 10 | 1958 | 89 | ||
BC034078.1 | - | 11 - 12 | 1645 | 89 | ||
BC076622.1 | - | 8 - 9 | 1020 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK154170.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
