Assay Details
Gene Symbol: Fam179b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177805.3 | NP_808473.2 | 3 - 4 | 2584 | 79 | |
GenBank mRNA | AK039995.1 | - | 4 - 5 | 2295 | 79 | |
AK141238.1 | - | 3 - 4 | 2524 | 79 | ||
AK172948.1 | - | 3 - 4 | 2512 | 79 | ||
BC157958.1 | - | 3 - 4 | 2386 | 9088131 | 79 | |
BC158008.1 | - | 3 - 4 | 2386 | 9007316 | 79 | |
BC172117.1 | - | 3 - 4 | 2386 | 9054403 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
