Assay Details
Gene Symbol: Pla2g4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK004701.2 | - | 4 - 5 | 544 | 122 | |
AK144750.1 | - | 5 - 6 | 501 | 122 | ||
BC003816.1 | - | 5 - 6 | 541 | 3485483 | 122 | |
M72394.1 | - | 5 - 6 | 489 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
