Assay Details
Gene Symbol: Baz1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013815.2 | NP_038843.2 | 16 - 17 | 2305 | 112 | |
GenBank mRNA | AK047427.1 | - | 1 - 2 | 112 | 112 | |
BC065123.1 | - | 2 - 3 | 121 | 112 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK047427.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
