Assay Details
Gene Symbol: Rasal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177644.5 | NP_808312.3 | 4 - 5 | 1219 | 75 | |
GenBank mRNA | AK039584.1 | - | 4 - 5 | 1219 | 75 | |
AK086812.1 | - | 4 - 5 | 845 | 75 | ||
BC120902.2 | - | 4 - 5 | 562 | 40086771 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
