Assay Details
Gene Symbol: Lct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081078.1 | NP_001074547.1 | 12 - 13 | 4890 | 101 | |
GenBank mRNA | AK158042.1 | - | 6 - 7 | 2992 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
