Assay Details
Gene Symbol: Camsap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081360.1 | NP_001074829.1 | 11 - 12 | 1871 | 82 | |
GenBank mRNA | AK028542.1 | - | 9 - 10 | 1821 | 82 | |
BC138367.1 | - | 10 - 11 | 1447 | 8861387 | 82 | |
BC145258.1 | - | 9 - 10 | 1414 | 9053811 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
