Assay Details
Gene Symbol: Camsap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081360.1 | NP_001074829.1 | 13 - 14 | 4134 | 83 | |
GenBank mRNA | AK028542.1 | - | 11 - 12 | 4084 | 83 | |
AK122436.1 | - | 1 - 2 | 1447 | 83 | ||
BC100407.1 | - | 1 - 2 | 1632 | 83 | ||
BC138367.1 | - | 12 - 13 | 3710 | 8861387 | 83 | |
BC145258.1 | - | 11 - 12 | 3677 | 9053811 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
