Assay Details
Gene Symbol: Camsap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081360.1 | NP_001074829.1 | 15 - 16 | 4484 | 66 | |
GenBank mRNA | AK081975.1 | - | 1 - 2 | 100 | 66 | |
BC138367.1 | - | 14 - 15 | 4060 | 8861387 | 66 | |
BC145258.1 | - | 13 - 14 | 4027 | 9053811 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
