Assay Details
Gene Symbol: Camsap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081360.1 | NP_001074829.1 | 7 - 8 | 1500 | 77 | |
GenBank mRNA | AK028542.1 | - | 5 - 6 | 1450 | 77 | |
BC138367.1 | - | 6 - 7 | 1076 | 8861387 | 77 | |
BC145258.1 | - | 5 - 6 | 1043 | 9053811 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
