Assay Details
Gene Symbol: Camsap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081360.1 | NP_001074829.1 | 9 - 10 | 1740 | 61 | |
GenBank mRNA | AK028542.1 | - | 7 - 8 | 1690 | 61 | |
BC138367.1 | - | 8 - 9 | 1316 | 8861387 | 61 | |
BC145258.1 | - | 7 - 8 | 1283 | 9053811 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
