Assay Details
Gene Symbol: Atp2b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167949.2 | NP_001161421.1 | 4 - 5 | 1244 | 62 | |
NM_213616.4 | NP_998781.1 | 4 - 5 | 1244 | 62 | ||
GenBank mRNA | AK143672.1 | - | 5 - 6 | 1365 | 62 | |
AY560895.1 | - | 4 - 5 | 778 | 62 | ||
BC109172.1 | - | 4 - 5 | 778 | 40048499 | 62 | |
BC109173.1 | - | 4 - 5 | 778 | 40048500 | 62 | |
EU493094.2 | - | 4 - 5 | 948 | 62 | ||
EU493095.2 | - | 4 - 5 | 948 | 62 | ||
JF683384.1 | - | 4 - 5 | 948 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
