Assay Details
Gene Symbol: Gm9696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_037189.1 | 1 - 2 | 126 | 158 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105227 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105227 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04922851 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105227 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175385 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175412 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175377 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175409 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175384 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175382 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175402 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175410 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02337283 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175422 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175414 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175391 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175418 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175419 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175424 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175387 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02426336 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175401 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03023608 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175395 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175397 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02337282 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175413 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175404 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175394 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175423 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175392 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03186332 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175426 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175376 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175400 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02669565 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22175396 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0300003318.mm.1 | TC0300000480.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300003318.mm.1 | NR_037189.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300003318.mm.1 | NR_037189.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300003317.mm.1 | TC0300000480.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300003317.mm.1 | NR_037189.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300003317.mm.1 | NR_037189.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300003319.mm.1 | TC0300000480.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300003319.mm.1 | NR_037189.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300003319.mm.1 | NR_037189.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300003316.mm.1 | TC0300000480.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300003316.mm.1 | NR_037189.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300003316.mm.1 | NR_037189.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300003320.mm.1 | TC0300000480.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300003320.mm.1 | NR_037189.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300003320.mm.1 | NR_037189.1 | Refseq |
Murine Genome U74v2 Set | 133984_PM_at | - | - |
Murine Genome U74v2 Set | 133984_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 411457 | - | - |
