Assay Details
Gene Symbol: Cntn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177129.5 | NP_796103.2 | 4 - 5 | 697 | 70 | |
GenBank mRNA | AK147602.1 | - | 4 - 5 | 698 | 70 | |
BC066106.1 | - | 4 - 5 | 617 | 30362931 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
