Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100027244.mm.1 |
TC0100003323.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100027244.mm.1 |
NM_001159965.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027244.mm.1 |
NM_001159965.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027244.mm.1 |
NM_001159966.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027244.mm.1 |
NM_001159966.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027244.mm.1 |
NM_001159967.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027244.mm.1 |
NM_001159967.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027244.mm.1 |
NM_023884.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027244.mm.1 |
NM_023884.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027244.mm.1 |
NR_027640.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027244.mm.1 |
NR_027640.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027245.mm.1 |
TC0100003323.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100027245.mm.1 |
NM_001159965.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027245.mm.1 |
NM_001159965.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027245.mm.1 |
NM_001159966.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027245.mm.1 |
NM_001159966.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027245.mm.1 |
NM_001159967.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027245.mm.1 |
NM_001159967.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027245.mm.1 |
NM_001159968.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027245.mm.1 |
NM_001159968.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027245.mm.1 |
NM_023884.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027245.mm.1 |
NM_023884.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027245.mm.1 |
NR_027640.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027245.mm.1 |
NR_027640.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027250.mm.1 |
TC0100003323.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100027250.mm.1 |
NM_001159965.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027250.mm.1 |
NM_001159965.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027250.mm.1 |
NM_001159967.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027250.mm.1 |
NM_001159967.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027250.mm.1 |
NM_001159968.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027250.mm.1 |
NM_001159968.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027250.mm.1 |
NM_023884.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027250.mm.1 |
NM_023884.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027250.mm.1 |
NR_027640.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027250.mm.1 |
NR_027640.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027249.mm.1 |
TC0100003323.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100027249.mm.1 |
NM_001159965.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027249.mm.1 |
NM_001159965.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027249.mm.1 |
NM_001159967.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027249.mm.1 |
NM_001159967.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027249.mm.1 |
NM_001159968.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027249.mm.1 |
NM_001159968.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027249.mm.1 |
NM_023884.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027249.mm.1 |
NM_023884.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027249.mm.1 |
NR_027640.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027249.mm.1 |
NR_027640.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027248.mm.1 |
TC0100003323.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100027248.mm.1 |
NM_001159966.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027248.mm.1 |
NM_001159966.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027246.mm.1 |
TC0100003323.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100027246.mm.1 |
NM_001159965.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027246.mm.1 |
NM_001159965.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027246.mm.1 |
NM_001159966.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027246.mm.1 |
NM_001159966.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027246.mm.1 |
NM_001159967.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027246.mm.1 |
NM_001159967.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027246.mm.1 |
NM_001159968.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027246.mm.1 |
NM_001159968.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027246.mm.1 |
NR_027640.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027246.mm.1 |
NR_027640.1 |
Refseq |