Assay Details
Gene Symbol: 4933439K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040558.1 | 1 - 2 | 292 | 110 | ||
GenBank mRNA | AK017125.1 | - | 1 - 2 | 294 | 110 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | DV053915.1 | - | 182 | Chr. - - [-] on Build | 110 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 37125 |
Gm37125 | -23.9kb | Anti-sense | Chr. 1 - 172512019 - 172518985 on NCBI Build GRCm38 | |
Downstream | V-set and immunoglobulin domain containing 8 | Vsig8 |
A030011M19, EG240916 |
5.1kb | Anti-sense | Chr. 1 - 172555938 - 172563717 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
