Assay Details
Gene Symbol: Usp9x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009481.2 | NP_033507.2 | 15 - 16 | 2368 | 95 | |
GenBank mRNA | AK049283.1 | - | 16 - 17 | 2481 | 95 | |
DQ086491.1 | - | 15 - 16 | 2329 | 95 | ||
U67874.1 | - | 15 - 16 | 2329 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
