Assay Details
Gene Symbol: E2f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007892.2 | NP_031918.2 | 4 - 5 | 664 | 73 | |
GenBank mRNA | AK156760.1 | - | 4 - 5 | 583 | 73 | |
BC003220.1 | - | 4 - 5 | 664 | 3482306 | 73 | |
X86925.1 | - | 4 - 5 | 530 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
